Mouse Recombinant MCP-5 (from E. coli)
Supplier: PeproTech
250-04-5UG 250-04-1MG 250-04-20UG 250-04-500UG 250-04-250UG 250-04-100UG
10773-134EA 150.18 CAD
10773-134 10773-144 10773-136 10773-142 10773-140 10773-138
Mouse Recombinant MCP-5 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Murine MCP-5 (CCL12) is a 9.3 kDa protein containing 82 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.