Mouse Recombinant Lungkine (from E. coli)
Supplier: PeproTech
250-37-100UG 250-37-5UG 250-37-500UG 250-37-1MG 250-37-20UG 250-37-250UG
10773-418EA 1126.37 CAD
10773-418 10773-414 10773-422 10773-424 10773-416 10773-420
Mouse Recombinant Lungkine (from E. coli)
Proteins and Peptides
Lungkine is a CXC chemokine that is expressed in lung epithelial cells and, to a lesser extent, in certain fetal tissues. No human homolog has been identified, and a specific cell surface receptor has not yet been found. Lungkine expression in lung tissue is elevated in response to inflammation, at which time it acts to specifically recruit neutrophils and direct them into the lung airway. Recombinant Murine Lungkine (CXCL15) is a 16.3 kDa CXC chemokine consisting of 142 amino acid residues, including four conserved cysteine residues present in CXC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.