Human Recombinant MIP-3 (from E. coli)
Supplier: PeproTech
300-29-100UG 300-29-1MG 300-29-20UG 300-29-500UG 300-29-5UG 300-29-250UG
10773-770EA 1126.37 CAD
10773-770 10773-776 10773-768 10773-774 10773-766 10773-772
Human Recombinant MIP-3 (from E. coli)
Proteins and Peptides
MIP-3 is a CC chemokine that signals through the CCR1 receptor. MIP-3 chemoattracts monocytes, resting T lymphocytes, and neutrophils, but does not chemoattract activated lymphocytes. Additionally, MIP-3 has been shown to inhibit colony formation of bone marrow myeloid immature progenitors. Recombinant Human MIP-3 (CCL23) is an 11.3 kDa protein containing 99 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.