Human Recombinant Exodus-2 (from E. coli)
Supplier: PeproTech
300-35-250UG 300-35-500UG 300-35-1MG 300-35-20UG 300-35-5UG 300-35-100UG 300-35-50UG
10773-866EA 1970.75 CAD
10773-866 10773-868 10773-870 10773-860 10773-858 10773-864 10773-862
Human Recombinant Exodus-2 (from E. coli)
Proteins and Peptides
Exodus-2 is a CC chemokine that signals through the CCR7 receptor. It is expressed in lymph nodes of certain endothelial cells, and in the spleen and appendix. Exodus-2 chemoattracts T and B lymphocytes and inhibits hematopoiesis. Exodus-2 contains six cysteine residues, including the four conserved cysteines present in CC chemokines. Recombinant Human Exodus-2 (CCL21) is a 12.2 kDa protein containing 111 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.