Human Recombinant gAcrp30/Adipolean (from E. coli)
Supplier: PeproTech
450-21-1MG 450-21-100UG 450-21-5UG 450-21-250UG 450-21-500UG 450-21-25UG
10774-650EA 5630.72 CAD
10774-650 10774-644 10774-640 10774-646 10774-648 10774-642
Human Recombinant gAcrp30/Adipolean (from E. coli)
Proteins and Peptides
gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver. Recombinant Human gAcrp30/Adipolean is a 16.6 kDa protein consisting of 145 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.