Human;Mouse Recombinant FGF-8b (from E. coli)
Supplier: PeproTech
AF10025100UG AF10025250UG AF1002525UG AF10025500UG AF100255UG AF100251MG
10778-062EA 1279.35 CAD
10778-062 10778-064 10778-060 10778-066 10778-058 10778-068
Human;Mouse Recombinant FGF-8b (from E. coli)
Proteins and Peptides
  • Animal-free
FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF-8A and B are identical, unlike human and mouse FGF-8E and F, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors. Recombinant Human/Murine FGF-8b is a 22.5 kDa protein consisting of 194 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.