Human Recombinant FGF-21 (from E. coli)
Supplier: PeproTech
AF10042100UG AF100425UG AF1004225UG AF100421MG AF10042500UG AF10042250UG
10778-136EA 1269.69 CAD
10778-136 10778-132 10778-134 10778-142 10778-140 10778-138
Human Recombinant FGF-21 (from E. coli)
Proteins and Peptides
  • Animal-free
FGF-21 is a secreted growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-21, in the presence of β-Klotho as a protein cofactor, signals through the FGFR 1c and 4 receptors, and stimulates insulin-independent glucose uptake by adipocytes. Recombinant Human FGF-21 is a 19.5 kDa protein containing 182 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.