Human Recombinant IL-33 (from E. coli)
Supplier: PeproTech
AF2003310UG AF200331MG AF20033100UG AF200332UG AF20033500UG AF20033250UG
10778-420EA 408.54 CAD
10778-420 10778-428 10778-422 10778-418 10778-426 10778-424
Human Recombinant IL-33 (from E. coli)
Proteins and Peptides
  • Animal-free
Human IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It binds and signals through the IL-1RL1/ST2 receptor, to activate NF-κB and MAP kinases. IL-33 induces production of TH2 cell related cytokines, including IL-4, IL-5 and IL-13, and exerts multiple inflammation related bioactivities. Recombinant Human IL-33 is a 17.9 kDa protein containing 159 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.