Mouse Recombinant IL-1beta (from E. coli)
Supplier: PeproTech
AF21111B500UG AF21111B250UG AF21111B1MG
10778-446EA 4959.38 CAD
10778-446 10778-444 10778-448
Mouse Recombinant IL-1beta (from E. coli)
Proteins and Peptides
  • Animal-free
IL-1β is a proinflammatory cytokine produced in a variety of cells, including monocytes, tissue macrophages, keratinocytes, and other epithelial cells. Both IL-1α and IL-1β bind to the same receptor and have similar, if not identical, biological properties. These cytokines have a broad range of activities including the stimulation of thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity, and the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. Recombinant Murine IL-1β is a 17.5 kDa protein containing 153 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.