Animal-Free Recombinant Murine TPO, PeproTech, Inc.
Supplier: Peprotech
AF315142UG AF31514100UG AF3151410UG AF3151450UG
10778-842EA 182.81 CAD
10778-842 10778-848 10778-844 10778-846
Animal-Free Recombinant Murine TPO, PeproTech, Inc.
Proteins and Peptides
  • Animal-free
TPO is a lineage-specific growth factor produced in the liver, kidney and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo . TPO signals through the c-mpl receptor, and acts as an important regulator of circulating platelets. Human and murine TPO exhibit cross-species reactivity. Recombinant Murine TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.