Human Cytokine
Supplier: PeproTech
HHSC3
10778-856EA 3846.21 CAD
10778-856
Human Cytokine
Proteins and Peptides
The Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
The package contains the following 4 vials:
100ug Human Flt3 Ligand
100ug Human SCF
100ug Human TPO
10ug Human IL-3
To request a CoA or MSDS for the components of the package, please Contact Us.
To request a CoA or MSDS for the components of the package, please Contact Us.
Carrier free.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.