Mouse;Rat Cytokine
Supplier: PeproTech
MHSC6
10778-862EA 3846.21 CAD
10778-862
Mouse;Rat Cytokine
Proteins and Peptides
The Murine Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo mouse hematopoietic stem cells.
The package contains the following 4 vials:
100ug Murine Flt3 Ligand
100ug Murine SCF
100ug Murine TPO
10ug Murine IL-6
To request a CoA or MSDS for the components of the package, please Contact Us.
Carrier free.
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.