To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Animal-free:Y
- Size:100 µg
- Species:Human
- Size:100 µg
- Storage conditions:−20 °C
- Endotoxin content:<0.1 ng/μg of protein (<1 EU/μg).
- Protein/peptide name:Cytokine
- Formulation:Lyophilized
- Shipping temperature:Room Temperature
- Cat. no.:10778-868
- Origin:USA
Specifications
About this item
The Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
- Animal-free
The package contains the following 4 vials:
100ug Animal-Free Human Flt3 Ligand
100ug Animal-Free Human SCF
100ug Animal-Free Human TPO
10ug Animal-Free Human IL-3
To request a CoA or MSDS for the components of the package, please Contact Us.
Manufactured using all Animal-Free reagents.
To request a CoA or MSDS for the components of the package, please Contact Us.