Mouse Recombinant RANTES (from E. coli)
Supplier: PeproTech
250-07-500UG 250-07-250UG 250-07-100UG 250-07-1MG
10780-236EA 3378.39 CAD
10780-236 10780-234 10780-232 10780-238
Mouse Recombinant RANTES (from E. coli)
Proteins and Peptides
RANTES is a CC chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant Murine RANTES (CCL5) is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.