Mouse Recombinant JE/MCP-1 (from E. coli)
Supplier: PeproTech
250-10-250UG 250-10-100UG 250-10-50UG 250-10-500UG 250-10-2UG 250-10-10UG
10780-276EA 1970.75 CAD
10780-276 10780-274 10780-272 10780-278 10780-268 10780-270
Mouse Recombinant JE/MCP-1 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Murine JE/MCP-1 (CCL2) is a 13.8 kDa protein containing 125 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.