Human Recombinant MCP-1 (from E. coli)
Supplier: PeproTech
300-04-5UG 300-04-1MG 300-04-250UG 300-04-500UG 300-04-20UG 300-04-100UG
10780-410EA 152.01 CAD
10780-410 10780-420 10780-416 10780-418 10780-412 10780-414
Human Recombinant MCP-1 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Human MCP-1 (CCL2) is an 8.6 kDa protein containing 76 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.