Human Recombinant RANTES (from E. coli)
Supplier: PeproTech
300-06-1MG 300-06-250UG 300-06-500UG 300-06-20UG 300-06-5UG 300-06-100UG 300-06-50UG
10780-446EA 5630.72 CAD
10780-446 10780-442 10780-444 10780-436 10780-434 10780-440 10780-438
Human Recombinant RANTES (from E. coli)
Proteins and Peptides
RANTES is a CC chemokine that can signal through the CCR1, CCR3, CCR5 and US28 (cytomegalovirus receptor) receptors. It is a chemoattractant towards monocytes, memory T cells (CD4+/CD45RO), basophils, and eosinophils. RANTES also has the capability to inhibit certain strains of HIV-1, HIV-2 and simian immunodeficiency virus (SIV). Recombinant Human RANTES (CCL5) is a 7.8 kDa protein containing 68 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.