Human Recombinant MCP-2 (from E. coli)
Supplier: PeproTech
300-15-10UG 300-15-500UG 300-15-1MG 300-15-250UG 300-15-2UG 300-15-100UG
10780-502EA 366.09 CAD
10780-502 10780-508 10780-510 10780-506 10780-500 10780-504
Human Recombinant MCP-2 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Human MCP-2 (CCL8) is an 8.9 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.