Rat Recombinant MCP-1 (from E. coli)
Supplier: PeproTech
400-12-250UG 400-12-1MG 400-12-2UG 400-12-10UG 400-12-100UG 400-12-500UG 400-12-50UG
10780-994EA 1970.75 CAD
10780-994 10780-998 10780-986 10780-988 10780-992 10780-996 10780-990
Rat Recombinant MCP-1 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Rat MCP-1 (CCL2) is a 14.0 kDa protein containing 125 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.