Rat Alarin Trifluoroacetate
Supplier: Bachem
4061788.1000 4061788.0500
H-6762.1000BAEA 525.41 CAD
H-6762.1000BA H-6762.0500BA
Rat Alarin Trifluoroacetate
Proteins and Peptides
Peptide belonging to the galanin family of peptides. Alarin exhibits potent dose-dependent vasoconstrictor and anti-edema activity in the cutaneous microvasculature. Alarin stimulated food intake and gonadotrophin release in male rats.