((D2)Gly4)-Cck-8 (Sulfated) Ammonium, Bachem
Supplier: BACHEM AMERICAS INC
4072027.1000
H-7248.1000BAEA 751.4 CAD
H-7248.1000BA
((D2)Gly4)-Cck-8 (Sulfated) Ammonium, Bachem
Proteins and Peptides
Heavy isotope-labeled sulfated CCK-8 useful for pharmacokinetic/pharmacodynamic studies.
Cholecystokinin (CCK) is a hormone originally isolated from porcine intestinal mucosa and described as a linear 33-amino acid peptide containing a sulfated tyrosine, which is essential for its biological activity. It has been found in mammals in both the digestive tract and the central nervous system. Among its multiple biological functions, this hormone stimulates pancreatic exocrine secretion, gallbladder contraction, and intestine motility and may also act as a neurotransmitter/neuromodulator in the central nervous system. For CCK-4 see H-3110.