((13C6)Leu5)-Ghrelin Trifluoroacetate, Bachem
Supplier: BACHEM AMERICAS INC
4072025.0500
H-7252.0500BAEA 1126.16 CAD
H-7252.0500BA
((13C6)Leu5)-Ghrelin Trifluoroacetate, Bachem
Proteins and Peptides
Stable isotope-labeled human ghrelin useful for pharmacokinetic/pharmacodynamic studies.