APL1B27 Trifluoroacetate
Supplier: Bachem
4074447.0500 4074447.1000
H-7304.0500BAEA 442.14 CAD
H-7304.0500BA H-7304.1000BA
APL1B27 Trifluoroacetate
Amyloid precursor-like protein 1 (APLP1) is a membrane-associated glycoprotein, whose gene is homologous to the APP gene. APLP1 was shown to be involved in the pathogenesis of Alzheimer's disease. Contrary to the β-amyloid peptides excised from APP, the peptides resulting from the enzymatic processing of APLP1 are not neurotoxic.