APL1B28 Trifluoroacetate
Supplier: Bachem
4074448.0500 4074448.1000
H-7306.0500BAEA 504.99 CAD
H-7306.0500BA H-7306.1000BA
APL1B28 Trifluoroacetate
Proteins and Peptides
A highly sensitive biomarker for the production of Aβ 1-42 derived from the βAPP-like protein APLP1. The peptide is generated by the same proteolytic mechanism as Aβ42. Non-amyloidogenic APL1β28 can be detected in the CSF and its levels correlate with Aβ42 production. Its ratio to total APL1β (APL1β25, APL1β27, and APL1β28) is significantly increased in familial and sporadic cases of AD.
Amyloid precursor-like protein 1 (APLP1) is a membrane-associated glycoprotein, whose gene is homologous to the APP gene. APLP1 was shown to be involved in the pathogenesis of Alzheimer's disease. Contrary to the β-amyloid peptides excised from APP, the peptides resulting from the enzymatic processing of APLP1 are not neurotoxic.