KODAK® GBX Developer and Fixer, Carestream Health
1901859 1900943 1901875 1900984
IB19009-43CS 736.55 CAD
GRKK1901859 IB19009-43 IB1901875 IB19009-84
KODAK® GBX Developer and Fixer, Carestream Health
GBX Developer, Fixer, and Replenisher are liquid chemical concentrates designed for the manual processing of dental X-ray films (both intraoral and extraoral).
  • Provides mixing convenience and improved chemical stability
  • Identical mixing and dilution procedures for tank and replenishment solutions
  • Both deveoper and fixer are single solution concentrates