Anti-CCL4 Rabbit Polyclonal Antibody [clone: C4HCLC]

Supplier: Pierce Biotechnology
710016
CAPI710016EA 1546.67 CAD
CAPI710016
Anti-CCL4 Rabbit Polyclonal Antibody [clone: C4HCLC]
This antibody is predicted to react with human based on sequence homology. ABfinity™ recombinant antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale and purified with Protein A. ABfinity™ oligoclonal antibodies comprise a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds—the sensitivity of a polyclonal antibody with the specificity of a monoclonal, all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody—recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets when compared with monoclonal antibodies—an oligoclonal antibody has a known mixture of light and heavy chains. This exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Macrophage inflammatory protein-1 beta is a chemokine belonging to the family of chemotactic cytokines. It is involved in chemotactic and proinflammatory effects, and homeostasis. It is produced by macrophages upon stimulation by bacterial endotoxins. It recruits and stimulates various inflammatory cells at sites of inflammation. MIP-1 beta is produced by lymphocytes, macrophages and dendritic cells. This protein binds with high affinity to CCR5 receptors.

Recommended Dilutions: Western Blot: 4-5 ?g/ml

Type: Primary
Antigen: CCL4
Clonality: Polyclonal
Clone: C4HCLC
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR