Anti-EGFR Rabbit Monoclonal Antibody [clone: 2H9L3]

Supplier: Pierce Biotechnology
701053
CAPI701053EA 1564.82 CAD
CAPI701053
Anti-EGFR Rabbit Monoclonal Antibody [clone: 2H9L3]
ABfinity™ recombinant antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale, and purified with Protein A. ABfinity™ monoclonal antibodies resemble rabbit monoclonals isolated from serum or produced by hybridomas, but demonstrate greater specificity and sensitivity. Because ABfinity™ recombinant antibodies are derived from cloned DNA sequences of the heavy and light antibody chains, they are not susceptible to cell-line drift or lot-to-lot variation, thus allowing for peak specificity and performance. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Epidermal Growth Factor Receptor (EGFR) is a 175 kDa transmembrane glycoprotein belonging to tyrosine kinase superfamily. It acts as a receptor for epidermal growth factor (EGF) family proteins. Binding of EGFR to its ligands causes autophosphorylation of tyrosine kinase followed by activation of signal transduction pathways connected to cell proliferation and differentiation. Tyrosine 1068 present within the cytoplasmic domain of the receptor is a major autophosphorylation site that allows binding of Grb2 and activation of the Ras-Raf-ERK1 and ERK2 signaling pathway.

Recommended Dilutions: Western Blot: 1:500-1:5000, Immunofluorescence: 1:100-1:1000, Immunocytochemistry: 1:100-1:1000, Flow Cytometry: 1:100-1:500

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 2H9L3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR