Anti-NAMPT Rabbit Monoclonal Antibody [clone: 13H13L23]

Supplier: Pierce Biotechnology
701090 701090
CAPI701090EA 1383.3 CAD
CAPI701090 10156-732
Anti-NAMPT Rabbit Monoclonal Antibody [clone: 13H13L23]
This antibody is predicted to react with mouse based on sequence homology. ABfinity™ recombinant antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale, and purified with Protein A. ABfinity™ monoclonal antibodies resemble rabbit monoclonals isolated from serum or produced by hybridomas, but demonstrate greater specificity and sensitivity. Because ABfinity™ recombinant antibodies are derived from cloned DNA sequences of the heavy and light antibody chains, they are not susceptible to cell-line drift or lot-to-lot variation, thus allowing for peak specificity and performance. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.

Recommended Dilutions: Western Blot: 2-5 ?g/ml, Immunofluorescence: 1-3 ?g/ml, Immunocytochemistry: 1-3 ?g/ml, Flow Cytometry: 2-4µg x 10^6 cells

Type: Primary
Antigen: NAMPT
Clonality: Monoclonal
Clone: 13H13L23
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR