Anti-GSK3B Rabbit Polyclonal Antibody [clone: 9HCLC]

Supplier: Pierce Biotechnology
710100
CAPI710100EA 1361.64 CAD
CAPI710100
Anti-GSK3B Rabbit Polyclonal Antibody [clone: 9HCLC]
This antibody is predicted to react with non-human primate, mouse and rat based on sequence homology. ABfinity™ recombinant antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale and purified with Protein A. ABfinity™ oligoclonal antibodies comprise a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds—the sensitivity of a polyclonal antibody with the specificity of a monoclonal, all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody—recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets when compared with monoclonal antibodies—an oligoclonal antibody has a known mixture of light and heavy chains. This exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Glycogen synthase kinase-3 beta (GSK3 beta) belongs to the subfamily of glycogen synthase kinases, that play a central role in energy metabolism, neuronal cell development, and body pattern formation. GSK3 has alpha and beta isoforms (51 kDa and 47 kDa respectively), that are functionally dissimilar. GSK3 beta plays a role in insulin and Wnt signaling, and is inhibited following the extracellular signaling events.

Recommended Dilutions: Western Blot: 1:500-1:5000, Immunofluorescence: 1:100-1:1000, Immunocytochemistry: 1:100-1:1000, ChIP assay: 1 ?l

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone: 9HCLC
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR