Anti-CDX1 Mouse Monoclonal Antibody [clone: 937CT11.6.2]

Supplier: Pierce Biotechnology
MA518113
CAPIMA518113EA 1857.5 CAD
CAPIMA518113
Anti-CDX1 Mouse Monoclonal Antibody [clone: 937CT11.6.2]
Could play a role in the terminal differentiation of the intestine.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CDX1
Clonality: Monoclonal
Clone: 937CT11.6.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR