Mouse IgG2a Isotype Control (Alexa Fluor® 647)

MA518170 MA518170
CAPIMA518170EA 624.47 CAD
10157-540 CAPIMA518170
Mouse IgG2a Isotype Control (Alexa Fluor® 647)
This product can be used as a negative control to measure the non-specific binding of Mouse IgG2a in human, porcine, mouse and rat tissues. The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody's species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Recommended Dilutions: Flow Cytometry: Assay-dependent, Control: Assay-dependent

Type: Control
Antigen: IgG2a
Clonality:
Clone: PPV-04
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR