Anti-DPP3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535038
CAPIPA535038EA 1774.17 CAD
CAPIPA535038
Anti-DPP3 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: DPP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR