Anti-EXOC3L Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535040
CAPIPA535040EA 1857.5 CAD
CAPIPA535040
Anti-EXOC3L Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. As part of the exocyst, may play a role in regulated exocytosis of insulin granules.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: EXOC3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR