Anti-PIP5KL1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535041
CAPIPA535041EA 1774.17 CAD
CAPIPA535041
Anti-PIP5KL1 Rabbit Polyclonal Antibody
PIP5KL1 is a phosphoinositide kinase-like protein that lacks intrinsic lipid kinase activity but associates with type I PIPKs (see PIP5K1A; MIM 603275) and may play a role in localization of PIPK activity.

PIP5KL1 is a phosphoinositide kinase-like protein that lacks intrinsic lipid kinase activity but associates with type I PIPKs and may play a role in localization of PIPK activity.

Type: Primary
Antigen: PIP5KL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR