Anti-ANKRD34B Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535050
CAPIPA535050EA 1774.17 CAD
CAPIPA535050
Anti-ANKRD34B Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: ANKRD34B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR