Anti-BTC Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535064
CAPIPA535064EA 1857.5 CAD
CAPIPA535064
Anti-BTC Rabbit Polyclonal Antibody
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: BTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR