Anti-TBX6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535065
CAPIPA535065EA 1960.37 CAD
CAPIPA535065
Anti-TBX6 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and rat based on sequence homology. This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Knockout studies in mice indicate that this gene is important for specification of paraxial mesoderm structures.

  • Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: TBX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR