Anti-LRP12 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535069
CAPIPA535069EA 1857.5 CAD
CAPIPA535069
Anti-LRP12 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and non-human primate based on sequence homology. This gene encodes a member of the low-density lipoprotein receptor related protein family. The product of this gene is a transmembrane protein that is differentially expressed in many cancer cells. Alternate splicing results in multiple transcript variants.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: LRP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR