Anti-SOS2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535070
CAPIPA535070EA 1825.53 CAD
CAPIPA535070
Anti-SOS2 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. SOS2 promotes the exchange of Ras-bound GDP by GTP.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SOS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR