Anti-TBC1D13 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535080
CAPIPA535080EA 1857.5 CAD
CAPIPA535080
Anti-TBC1D13 Rabbit Polyclonal Antibody
TBC1D13 may act as a GTPase-activating protein for Rab family protein(s).

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100

Type: Primary
Antigen: TBC1D13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR