Anti-METTL2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535085
CAPIPA535085EA 1857.5 CAD
CAPIPA535085
Anti-METTL2 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and zebrafish based on sequence homology. METTL2A is probable methyltransferase.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100

Type: Primary
Antigen: METTL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR