Anti-AP1M1 Rabbit Polyclonal Antibody

PA535098 PA535098
CAPIPA535098EA 934.8 CAD
CAPIPA535098 10157-708
Anti-AP1M1 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. The protein encoded by this gene is the medium chain of the trans-Golgi network clathrin-associated protein complex AP-1. The other components of this complex are beta-prime-adaptin, gamma-adaptin, and the small chain AP1S1. This complex is located at the Golgi vesicle and links clathrin to receptors in coated vesicles. These vesicles are involved in endocytosis and Golgi processing. Alternatively spliced transcript variants encoding distinct protein isoforms have been found for this gene.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: AP1M1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR