Anti-RBPJL Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535099
CAPIPA535099EA 2043.02 CAD
CAPIPA535099
Anti-RBPJL Rabbit Polyclonal Antibody
In mouse, recombining binding protein L (RBP-L) is a transcription factor that binds to DNA sequences almost identical to that bound by the Notch receptor signalling pathway transcription factor RBP-J. However, unlike RBP-J, RBP-L does not interact with Notch receptors. RBP-L has been shown to activate transcription in concert with Epstein-Barr virus nuclear antigen-2 (EBNA2). The protein encoded by this gene is similar in sequence to the mouse RPB-L protein and Drosophila suppressor of hairless protein.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: RBPJL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR