Anti-CACNG6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535100
CAPIPA535100EA 1857.5 CAD
CAPIPA535100
Anti-CACNG6 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. Voltage-dependent calcium channels are composed of five subunits. The protein encoded by this gene represents one of these subunits, gamma, and is one of two known gamma subunit proteins. This particular gamma subunit is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state. This gene is part of a functionally diverse eight-member protein subfamily of the PMP-22/EMP/MP20 family and is located in a cluster with two family members that function as transmembrane AMPA receptor regulatory proteins (TARPs). Alternative splicing results in multiple transcript variants. Variants in this gene have been associated with aspirin-intolerant asthma.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: CACNG6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR