Anti-TBPL2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535109
CAPIPA535109EA 1857.5 CAD
CAPIPA535109
Anti-TBPL2 Rabbit Polyclonal Antibody
TBPL2 is a transcription factor required in complex with TAF3 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: TBPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR