Anti-DNAJB11 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535122
CAPIPA535122EA 1857.5 CAD
CAPIPA535122
Anti-DNAJB11 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. DNAJB11 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity bystimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a C-terminal cysteine-rich region.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: DNAJB11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR