Anti-HINFP Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535123
CAPIPA535123EA 1960.37 CAD
CAPIPA535123
Anti-HINFP Rabbit Polyclonal Antibody
MIZF interacts with methyl-CpG-binding protein-2 (MBD2; MIM 603547), a component of the MeCP1 histone deacetylase (HDAC) complex, and plays a role in DNA methylation and transcription repression.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HINFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR