Anti-LZTS1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535130
CAPIPA535130EA 1857.5 CAD
CAPIPA535130
Anti-LZTS1 Rabbit Polyclonal Antibody
This gene encodes a tumor suppressor protein that is ubiquitously expressed in normal tissues. In uveal melanomas, expression of this protein is silenced in rapidly metastasizing and metastatic tumor cells but has normal expression in slowly metastasizing or nonmetastasizing tumor cells. This protein may have a role in cell-cycle control by interacting with the Cdk1/cyclinB1 complex. This gene is located on chromosomal region 8p22. Loss of heterozygosity (LOH) in the 8p arm is a common characteristic of many types of cancer.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: LZTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR