Anti-CSRNP2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535134
CAPIPA535134EA 1857.5 CAD
CAPIPA535134
Anti-CSRNP2 Rabbit Polyclonal Antibody
CSRNP2 binds to the consensus sequence 5'-AGAGTG-3' and has transcriptional activator activity. May play a role in apoptosis.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: CSRNP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR