Anti-TERF2IP Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA535137
CAPIPA535137EA 1774.17 CAD
CAPIPA535137
Anti-TERF2IP Rabbit Polyclonal Antibody
This antibody is predicted to react with non-human primate based on sequence homology. The gene encodes a protein that is part of a complex involved in telomere length regulation. Pseudogenes are present on chromosomes 5 and 22.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10-1:50

Type: Primary
Antigen: TERF2IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR